Learn More
BIOHIPPO Carboxyethyl-Cysteine pAb
SDP
Catalog No. 5000975470
Encompass
U-Blot Pan-Carboxyethyl-Cysteine Rabbit pAb is a Rabbit-derived polyclonal antibody that specifically recognizes Carboxyethyl-Cysteine a cysteine carboxyethylation modification at Cys(C) It shows reactivity with Species Independent Validated applications include WB IHC-P ICC/IF FC Recommended dilutions WB 11000 IHC-P 1100-1200 ICC/IF 1100-1200 FC 1100-1200 Purified by protein a Carboxyethyl-Cysteine (CEC) is a novel post-translational modification occurring on cysteine residues of proteins This modification is formed by covalently attaching a carboxyethyl group to the thiol group (-SH) of cysteine generating a stable thioether linkage Supplied as 100 uL
Catalog No. 50-009-75470 Supplier BIOHIPPO Supplier No. BHA21821315
U-Blot Pan-Carboxyethyl-Cysteine Rabbit pAb is a Rabbit-derived polyclonal antibody that specifically recognizes Carboxyethyl-Cysteine a cysteine carboxyethylation modification at Cys(C) It shows reactivity with Species Independent Validated applications include WB IHC-P ICC/IF FC Recommended dilutions WB 11000 IHC-P 1100-1200 ICC/IF 1100-1200 FC 1100-1200 Purified by protein a Carboxyethyl-Cysteine (CEC) is a novel post-translational modification occurring on cysteine residues of proteins This modification is formed by covalently attaching a carboxyethyl group to the thiol group (-SH) of cysteine generating a stable thioether linkage Supplied as 100 uL