Learn More
Abcam 5(6)-Carbo x yfluorescein, NHS ester, 25MG
Supplier:  Abcam AB27536525MG
Encompass_Preferred
MW 473.4 g/mol, Purity >90%. Fluorescent, amine-reactive probe for bioconjugate synthesis. Labels bioconjugates on nucleophilic amines (N-terminus, lysine residues). Labeled bioconjugates resistant to hydrolysis. High stability and performance at physiological pH.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-186