Learn More
Bioworld CARBOXYL COUPLING KIT
Supplier:  Bioworld 201500011
Encompass_Preferred
bio-World's Carboxy Coupling Kitconsists of amino-terminated Separopore 4B-CL (4% crosslinked agarose beads). This kit is suitablefor covalent immobilization of peptides or other carboxyl-containing (COOH) molecules for use in affinity p...
Catalog No. 50-197-557