Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carboxyl diazirine alkyne | 1450754-37-6 | 98.0% | 166.18 | C8H10N2O2 | 25 MG
SDP

Catalog No. 5000209487
Encompass_Preferred

Carboxyl diazirine alkyne is a photoaffinity labeling reagent containing a diazirine photoreactive group and an alkyne handle for click chemistry. It is used for protein cross-linking, target identification, and inhibition and labeling studies of FTO and related demethylases.

  • Contains diazirine photoreactive group for UV-activated covalent cross-linking.
  • Alkyne handle enables copper-catalyzed click conjugation for downstream analysis.
  • High purity (98.0%) suitable for biochemical assays.
  • Supplied in small research quantities, including 25 mg packs.
  • Storage: pure form -20°C; in solvent -80°C for long-term stability.
  • Appearance: off-white to light yellow solid-liquid mixture.

Catalog No. 50-002-09487 Supplier Medchemexpress LLC Supplier No. 25MGHYW074785
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carboxyl diazirine alkyne is a photoaffinity labeling reagent containing a diazirine photoreactive group and an alkyne handle for click chemistry. It is used for protein cross-linking, target identification, and inhibition and labeling studies of FTO and related demethylases.

  • Contains diazirine photoreactive group for UV-activated covalent cross-linking.
  • Alkyne handle enables copper-catalyzed click conjugation for downstream analysis.
  • High purity (98.0%) suitable for biochemical assays.
  • Supplied in small research quantities, including 25 mg packs.
  • Storage: pure form -20°C; in solvent -80°C for long-term stability.
  • Appearance: off-white to light yellow solid-liquid mixture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.