Learn More
Spherotech CARBOXYL FLUORESCENT PARTICLES
Catalog No. NC2155880
Encompass_Preferred
Carboxyl Fluorescent Particles, Pink, 1% w/v, 0.04-0.08 um, 2 mL
Catalog No. NC2155880 Supplier Spherotech Supplier No. CFP005582
Carboxyl Fluorescent Particles, Pink, 1% w/v, 0.04-0.08 um, 2 mL