Learn More
Electron Microscopy Sciences Aurion Carboxyl Functionalized Gold Nanoparticles, 15 nm 5 ML
SDP
Supplier:  Electron Microscopy Sciences 25602
Encompass_Preferred
This product has been developed to facilitate conjugating gold nanoparticles to molecules that cannot be conjugated via the classic direct adsorption method. The product is especially suited for covalent conjugation of small ligands. Aurion Gold Nanoparticles (Carboxyl Functionalized) are polyethylene glycol-carboxyl stabilized gold nanoparticle solutions based on defined particle sizes of 6, 10, 15 and 25nm.
Particle size: 15 nm
Catalog No. 50-193-1086