Learn More
Polysciences, Inc. Polybead Carboxylate Microspheres 1.00m | 15mL
SDP
Catalog No. NC1343365
Encompass
Polybead Carboxylate Microspheres are monodisperse polystyrene microspheres that contain surface carboxyl groups. This allows for the covalent binding of proteins to the bead surface. These microspheres are packaged as 2.5% solids (w/v) aqueous suspensions.
Catalog No. NC1343365 Supplier Polysciences, Inc. Supplier No. 0822615
Polybead Carboxylate Microspheres are monodisperse polystyrene microspheres that contain surface carboxyl groups. This allows for the covalent binding of proteins to the bead surface. These microspheres are packaged as 2.5% solids (w/v) aqueous suspensions.