Learn More
Sigma Aldrich Fine Chemicals Biosciences Carboxylesterase 1 isoform
Catalog No. 502904292
Encompass
Carboxylesterase 1 is a member of a large multigene carboxylesterase family. These enzymes are responsible for the hydrolysis of ester- and amide-bond-containing drugs such as cocaine and heroin. They also hydrolyze long-chain fatty acid esters and thioesters. This enzyme is known to hydrolyze aromatic and aliphatic esters and is necessary for cellular cholesterol esterification. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds.
Catalog No. 50-290-4292 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E02871VL
Carboxylesterase 1 is a member of a large multigene carboxylesterase family. These enzymes are responsible for the hydrolysis of ester- and amide-bond-containing drugs such as cocaine and heroin. They also hydrolyze long-chain fatty acid esters and thioesters. This enzyme is known to hydrolyze aromatic and aliphatic esters and is necessary for cellular cholesterol esterification. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds.