Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carboxylesterase-IN-3 | 2764748-92-5 | 98.3% | C11H6Cl2N4OS | 10MG
SDP

Catalog No. 5000259289
Encompass_Preferred

Carboxylesterase-IN-3 is a potent Notum (carboxylesterase) inhibitor supplied as a high-purity research compound for in vitro pharmacology and biochemical assays. It exhibits reported IC50 ≤ 10 nM and is provided as a solid powder with documented solubility and storage conditions for assay preparation (CAS 2764748-92-5; formula C11H6Cl2N4OS; Mw 313.16).

  • High purity: 98.3%.
  • Potent Notum (carboxylesterase) inhibition, IC50 ≤ 10 nM.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance may be required.
  • Supplied as a powder suitable for formulation into assay solutions.
  • Recommended storage: powder at -20°C for long-term stability.
  • Available in multiple milligram-scale pack sizes for research use.

Catalog No. 50-002-59289 Supplier Medchemexpress LLC Supplier No. HY14268910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carboxylesterase-IN-3 is a potent Notum (carboxylesterase) inhibitor supplied as a high-purity research compound for in vitro pharmacology and biochemical assays. It exhibits reported IC50 ≤ 10 nM and is provided as a solid powder with documented solubility and storage conditions for assay preparation (CAS 2764748-92-5; formula C11H6Cl2N4OS; Mw 313.16).

  • High purity: 98.3%.
  • Potent Notum (carboxylesterase) inhibition, IC50 ≤ 10 nM.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance may be required.
  • Supplied as a powder suitable for formulation into assay solutions.
  • Recommended storage: powder at -20°C for long-term stability.
  • Available in multiple milligram-scale pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.