Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carboxylesterase-IN-3 | 2764748-92-5 | MFCD34676411 | 98.3% | 313.16 | C11H6Cl2N4OS | 5 MG
SDP

Catalog No. 5000221699
Encompass_Preferred

Carboxylesterase-IN-3 is a research-grade small molecule inhibitor of the carboxylesterase Notum enzyme, reported as compound 4y with an IC50 ≤ 10 nM. It is intended for biochemical and cellular studies of Wnt signaling and related disease research.

  • Potent Notum inhibitor (IC50 ≤ 10 nM).
  • High purity, 98.3%.
  • Molecular weight 313.16 g/mol; formula C11H6Cl2N4OS.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Available in multiple milligram pack sizes and as DMSO solutions.
  • Powder storage: -20°C for long term; in solvent: -80°C recommended.

Catalog No. 50-002-21699 Supplier Medchemexpress LLC Supplier No. HY1426895MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carboxylesterase-IN-3 is a research-grade small molecule inhibitor of the carboxylesterase Notum enzyme, reported as compound 4y with an IC50 ≤ 10 nM. It is intended for biochemical and cellular studies of Wnt signaling and related disease research.

  • Potent Notum inhibitor (IC50 ≤ 10 nM).
  • High purity, 98.3%.
  • Molecular weight 313.16 g/mol; formula C11H6Cl2N4OS.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Available in multiple milligram pack sizes and as DMSO solutions.
  • Powder storage: -20°C for long term; in solvent: -80°C recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.