Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chiron Carboxylic Acid and Salt Standards
SDP

Catalog No. 113671350ME Shop All Spex CertiPrep Products
Change view
Click to view available options
Chemical Name or Material:
Perfluoroethanoic acid
Perfluorotridecanoic acid, n-isomer (major)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
11-367-1350ME Perfluorotridecanoic acid, n-isomer (major)
15-118-250ME Perfluoroethanoic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-367-1350ME Supplier Chiron Supplier No. 11367.1350ME

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material Perfluorotridecanoic acid, n-isomer (major)
CAS 72629-94-8
Concentration 50 μg
Color Colorless
Synonym PFTrDA
Type Reference Material
Solvent or Matrix Methanol
Industry Type PFA Testing
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.