Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CarboxyLink™ Immobilization Kit, 2 mL

Catalog No. PI44899 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Column Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI44899 5 Column Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI44899 Supplier Thermo Scientific™ Supplier No. 44899
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Covalently immobilize peptide antigens and other carboxylate ligands for affinity purification using DADPA-activated agarose beads and EDC crosslinker.

Thermo Scientific™ CarboxyLink Coupling Resins and Kits provide for the covalent immobilization of peptides or other carboxyl-containing (-COOH) molecules to a porous, beaded resin for use in affinity purification procedures.

These activated resins enable immobilization of carboxylate ligands for use in affinity protocols. CarboxyLink Resin is crosslinked beaded agarose (or acrylamide-based Thermo Scientific UltraLink™ Biosupport) that has been activated with diaminodipropylamine (DADPA) to contain long spacer arms, each of which have a primary amine at the end. When incubated with the resin and the carbodiimide crosslinker EDC (included in the CarboxyLink Immobilization Kit), carboxyl-containing molecules become permanently attached to the support by stable amide bonds. Once a molecule is coupled, the prepared affinity column can be used multiple times in typical protein affinity purification procedures. CarboxyLink Coupling Resins also can be used to immobilize other kinds of molecules using alternative amine-reactive crosslinking chemistries.

Highlights:

  • CarboxyLink Coupling Resins – beaded agarose or acrylamide support activated with diaminodipropylamine (DADPA) for amine-to-carboxyl conjugation using EDC crosslinker

  • Efficient peptide immobilization – easily couple 1 to 2mg of peptide per milliliter of settled resin

  • Stable linkage – immobilization results in covalent attachment of carboxyl groups by amide bonds, allowing for multiple rounds of affinity purification with one batch of prepared resin

  • Flexible and gentle coupling conditions – immobilization reaction completed in simple MES or other non-amine and non-carboxyl, near-neutral buffer, with or without organic solvent

  • Ideal for unmodified peptides – immobilizes peptides with high capacity and various orientations without steric hindrance, allowing for effective use in affinity purification of specific antibodies

  • Convenient kits and product sizes – choose 5-column kits with complete sets of buffers, crosslinker and versatile spin/drip columns containing either type of resin (agarose or polyacrylamide) or choose stand-alone resin for other uses

CarboxyLink Coupling Resin (Part No. 20266, 44899):

  • Ligand: diaminodipropylamine (DADPA)
  • Support: crosslinked 4% beaded agarose
  • Spacer arm: 9 atoms
  • Loading: activated with at least 16μmol amine per milliliter of settled resin
  • Format: 50% resin slurry in 0.05% sodium azide

UltraLink DADPA Resin (Part No. 53154):

  • Ligand: diaminodipropylamine (DADPA)
  • Support: Thermo Scientific UltraLink Biosupport
  • Spacer arm: 14 atoms
  • Loading: activated with at least 40μmol amine per milliliter of settled resin
  • Format: 50% resin slurry in 0.02% sodium azide

Includes:

CarboxyLink Resin Columns (DADPA agarose or UltraLink Support), EDC crosslinker, coupling buffer (MES-buffered saline), wash solution, and column accessories

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 mL
Description CarboxyLink Immobilization Kit, 2mL
Content And Storage Sufficient For: Immobilizing 1 to 2 mg of peptide per column
• CarboxyLink Columns, 2 mL, 5 columns
• EDC Crosslinker, 5 x 60 mg
• MES Buffered Saline Pack (makes 500 mL), 1 pack
• Wash Solution, 120 mL
• Column Accessories

Store EDC desiccated at -20°C. Store other components at 4°C.
Product Line CarboxyLink
Quantity 5 Column Kit
Target Antibody
Type Immobilization Kit
Stationary Phase Amine
Sufficient For 5 Columns
Format Spin Column
Column Type Affinity, Agarose Resin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.