Learn More
AdipoGen Carboxymethyl-fluoresceinamide
SDP
Supplier:  AdipoGen CDXD0201M125
Encompass_Preferred
Chemical. CAS 442151-50-0. Formula C24H19NO6. MW 417.41. Synthetic. BioReagent, suitable for fluorescence.
Catalog No. 50-256-1191