Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Carboxypeptidase B, Porcine Pancreas

Catalog No. 2173561MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-735-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-735-61MG Supplier MilliporeSigma™ Supplier No. 2173561MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native carboxypeptidase B catalyzes the release of C-terminal from basic amino acids, arginine, lysine, or ornithine from polypeptides

  • Inhibited by EDTA and other Zn2+ chelators
  • Following reconstution aliquot and freeze. Stock Solutions are stable for up to 6 months at 20°C
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 360μM HEPES, 1.41μM zinc chloride, 1.41mg/mL Trehalose, pH 7.4
pH Range Optimum 7.0 to 9.0
CAS 9025-24-5
Content And Storage -20°C
Form Lyophilized
Quantity 1 mg
Solubility Water
Product Type Carboxypeptidase B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.