Learn More
Medchemexpress LLC Carboxypeptidase B1/ 100ug | 100ug
SDP
Catalog No. 5000186842
Encompass_Preferred
Carboxypeptidase B1/CPB1 Protein Human (HEK293 His) is an approximately 47 0 kDa carboxypeptidase B1 protein with a C-terminal His-flag
Catalog No. 50-001-86842 Supplier Medchemexpress LLC Supplier No. HYP7734100UG
Carboxypeptidase B1/CPB1 Protein Human (HEK293 His) is an approximately 47 0 kDa carboxypeptidase B1 protein with a C-terminal His-flag