Learn More
Medchemexpress LLC Carboxypeptidase B1/ 100ug
SDP
Catalog No. 5000181870
Encompass_Preferred
Carboxypeptidase B1/CPB1 protein is part of the PDGF/VEGF growth factor family and has the same structural and functional characteristics as proteins that affect cell growth proliferation and angiogenesis This family is critical for mediating endothelial cell responses and promoting blood vessel formation including VEGF-C which is involved in lymphangiogenesis Carboxypeptidase B1/CPB1 Protein Mouse (HEK293 His) is the recombinant mouse-derived Carboxypeptidase B1/CPB1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81870 Supplier Medchemexpress LLC Supplier No. HYP72869100UG
Add to Cart
Add to Cart
Carboxypeptidase B1/CPB1 protein is part of the PDGF/VEGF growth factor family and has the same structural and functional characteristics as proteins that affect cell growth proliferation and angiogenesis This family is critical for mediating endothelial cell responses and promoting blood vessel formation including VEGF-C which is involved in lymphangiogenesis Carboxypeptidase B1/CPB1 Protein Mouse (HEK293 His) is the recombinant mouse-derived Carboxypeptidase B1/CPB1 protein expressed by HEK293 with C-His labeled tag