Learn More
Cayman Chemical CarboxyphosphamIde 10mg
SDP
Catalog No. 502946934
Encompass_Preferred
An inactive metabolite of cyclophosphamide
Catalog No. 50-294-6934 Supplier Cayman Chemical Supplier No. 3405210 MG
An inactive metabolite of cyclophosphamide