Learn More
Cayman Chemical Carboxy-PTIO (potassium salt), Purity: ≥99%, CAS: 148819-94-7, 100 mg
SDP
Catalog No. 502893455
Encompass_Preferred
Carboxy-PTIO is a NO scavenger. It reacts stoichiometrically with NO and can be used for EPR detection of NO.{1203,4117}
Catalog No. 50-289-3455 Supplier Cayman Chemical Supplier No. 81540100 MG
Carboxy-PTIO is a NO scavenger. It reacts stoichiometrically with NO and can be used for EPR detection of NO.{1203,4117}