Learn More
Abcam 5(6)-Carbo x yrhodamine 110, HCl salt, 50MG
Supplier:  Abcam AB27592250MG
Encompass_Preferred
MW 374.3 Da, Purity >95%. Stable, long wavelength, water soluble fluorophore suitable for DNA/protein labeling and cell staining. Despite having a rhodamine nucleus, this probe has excitation and emission wavelengths similar to fluorescein (green color fluorescence), making it useful in detection systems employing fluorescein filter sets.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-131