Learn More
Abcam 3-Carbo x yumbelliferyl ß-D-galactoside, BSA conjugate, 2MG
Catalog No. 50996126
Encompass_Preferred
MW 66800 g/mol. A conjugate of BSA (bovine serum albumin, MW = 66.4K) and the beta-galactosidase substrate carboxyumbelliferyl beta-D-galactopyranoside ab275363 produces a protein that can be used to measure extracellular β-galactosidase activity, measure lacY permease activity, for microinjection analyses, or measure cell disruption or lysis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-126 Supplier Abcam Supplier No. AB2753632MG
MW 66800 g/mol. A conjugate of BSA (bovine serum albumin, MW = 66.4K) and the beta-galactosidase substrate carboxyumbelliferyl beta-D-galactopyranoside ab275363 produces a protein that can be used to measure extracellular β-galactosidase activity, measure lacY permease activity, for microinjection analyses, or measure cell disruption or lysis.
The product is subject to the following: Abcam Restricted Use Statement