Learn More
Abcam Carbo x yumbelliferyl ß-D-glucuronide (CUGlcU), 10MG
Catalog No. 50996091
Encompass_Preferred
MW 382.27 Da, Purity >95%. Water soluble fluorogenic GUS substrate similar to MUGlcU. Useful for measuring cloned β-glucuronidase activity in plant lysate, homogenate, leaf disk or whole plant assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-091 Supplier Abcam Supplier No. AB27507310MG
MW 382.27 Da, Purity >95%. Water soluble fluorogenic GUS substrate similar to MUGlcU. Useful for measuring cloned β-glucuronidase activity in plant lysate, homogenate, leaf disk or whole plant assays.
The product is subject to the following: Abcam Restricted Use Statement