Learn More
Foxx Life Sciences EZBio® 10L (2.5 GAL) Gamma Sterilized Double Bagged Autoclavable Light Blocking Dark Amber Polypropylene (PP) Carboy with 83mm (83B) VersaCap® Closure
GREENER_CHOICE
Catalog No. 501720665
Encompass_Preferred
Foxx Life Sciences EZBio® Gamma Sterile Carboys provide users with a highly versatile container that maximizes storage efficiency & ease of use. Assemblies are made in entirely with USP Class VI components & Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the carboy & molded grips in the base provide a full, secure grip to make lifting, handling, & pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material & volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.Dark Amber Polypropylene carboys are made from a USP Class VI, FDA Grade, autoclavable material.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling & cleaning.VersaCap® offers a wide variety of top connection options. 13L Max Capactiy.
Catalog No. 50-172-0665 Supplier Foxx Life Sciences Supplier No. 51N5G51FLS
Foxx Life Sciences EZBio® Gamma Sterile Carboys provide users with a highly versatile container that maximizes storage efficiency & ease of use. Assemblies are made in entirely with USP Class VI components & Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the carboy & molded grips in the base provide a full, secure grip to make lifting, handling, & pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material & volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.Dark Amber Polypropylene carboys are made from a USP Class VI, FDA Grade, autoclavable material.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling & cleaning.VersaCap® offers a wide variety of top connection options. 13L Max Capactiy.