Learn More
Cole-Parmer Cole-Parmer Narrow Mouth Carboy, Heavy Walled HDPE, 10L
Supplier:  Cole-Parmer 0603595
Encompass_Preferred
Economical, durable carboys are resistant to most chemicals. Heavy-duty swing handle allows easy carrying for your liquids and powders. Narrow-Mouth Carboys are complete with rubber O-ring for added security. HDPE closure contains tag holes for identification.
Catalog No. NC1387111