Learn More
Foxx Life Sciences EZBio® 2.5L (0.5 GAL) Gamma Sterilized Double Bagged HDPE Carboy with 83mm (83B) VersaCap® Closure
Catalog No. 501720669
Encompass_Preferred
Foxx Life Sciences EZBio® Gamma Sterile HDPE Carboys provide users with a highly versatile container that maximizes storage efficiency and ease of use. Carboys are made entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Large handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.HDPE carboys are made from a USP Class VI, FDA Grade materials.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options. 3.25L Max Capacity.
Catalog No. 50-172-0669 Supplier Foxx Life Sciences Supplier No. 5125C51FLS
Foxx Life Sciences EZBio® Gamma Sterile HDPE Carboys provide users with a highly versatile container that maximizes storage efficiency and ease of use. Carboys are made entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Large handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.HDPE carboys are made from a USP Class VI, FDA Grade materials.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options. 3.25L Max Capacity.