Learn More
Foxx Life Sciences EZclear 20L (5 Gal) Polycarbonate (PC) Carboy with Wide Mouth 120mm VersaCap and 26.5L Max Capacity, Large Pinched Handles
Catalog No. NC1818229
Encompass_Preferred
The EZclear Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows the user to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.PC carboys are made from a USP Class VI, FDA Grade materials. Rigid and clear material that can withstand three autoclaving cycles.
Catalog No. NC1818229 Supplier Foxx Life Sciences Supplier No. 1583121OEM
The EZclear Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows the user to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.PC carboys are made from a USP Class VI, FDA Grade materials. Rigid and clear material that can withstand three autoclaving cycles.