Learn More
Bellco Glass, Inc. Carboy, 20L, PETG, 120mm Cap, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800344
Encompass_Preferred
The EZclear Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. The optional wide neck allows the user to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.PETG carboys are made from a USP Class VI, FDA Grade, BPA-Free materials. Height (w/Cap): 25.2" (640mm)Height (w/o Cap) 23.8" (605mm)Length: 13.58" (345mm)Width: 8.07" (205mm)Approximate Capacity(max): 26.5LCarboy Material: PETGCap Material: Polypropylene
Catalog No. 50-255-0418