Learn More
Foxx Life Sciences Wide Mouth 20L Round Polypropylene (PP) Carboy with White Cap
Catalog No. NC1818186
Encompass_Preferred
Foxx Life Sciences offers a premium line of high quality, Round Wide Mouth Polypropylene (PP) Laboratory Carboys. Our versatile carboys with strong easy grip handles aim to take full advantage of storage. The Wide Mouth carboys are leak proof and allows for easy cleaning efficiency. Overall functionality is available in 10 Liter or 20 Liter sizes, without spigots each with a White HDPE material Cap. The Wide Mouth Carboy are Autoclavable. Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).Cap should be removed before Autoclaving this carboy.
Catalog No. NC1818186 Supplier Foxx Life Sciences Supplier No. 155C187FLS
Foxx Life Sciences offers a premium line of high quality, Round Wide Mouth Polypropylene (PP) Laboratory Carboys. Our versatile carboys with strong easy grip handles aim to take full advantage of storage. The Wide Mouth carboys are leak proof and allows for easy cleaning efficiency. Overall functionality is available in 10 Liter or 20 Liter sizes, without spigots each with a White HDPE material Cap. The Wide Mouth Carboy are Autoclavable. Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).Cap should be removed before Autoclaving this carboy.