Learn More
Foxx Life Sciences EZBio® 40L (10 GAL) Gamma Sterilized Double Bagged HDPE Wide Mouth Carboy with 120mm VersaCap® Closure
GREENER_CHOICE
Catalog No. 501720674
Encompass_Preferred
Foxx Life Sciences EZBio® Gamma Sterile HDPE Carboys provide users with a highly versatile container that maximizes storage efficiency and ease of use. Carboys are made in entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Large handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.HDPE carboys are made from a USP Class VI, FDA Grade materials.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options. 50L Max Capacity.
Catalog No. 50-172-0674 Supplier Foxx Life Sciences Supplier No. 5125J61FLS
Foxx Life Sciences EZBio® Gamma Sterile HDPE Carboys provide users with a highly versatile container that maximizes storage efficiency and ease of use. Carboys are made in entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Large handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.HDPE carboys are made from a USP Class VI, FDA Grade materials.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options. 50L Max Capacity.