Learn More
Grainger Carboy: Blow Molded Carboy/Jerrican/Jug Handle, 5 gal Labware Capacity - English, HDPE
Catalog No. NC0300511
Encompass_Preferred
This lightweight carboy from Dynalon Labware has large carrying handles and indented hand grips on the bottom for easy carrying and pouring. The high-density polyethylene (HDPE) material has high impact strength and excellent compatibility with weak and concentrated acids, bases, and alcohol. Supplied with a 63mm threaded closure that can be drilled to accept a faucet. Overall Height 342 mm, Overall Depth 245 mm, Overall Width248 mm
Catalog No. NC0300511 Supplier Grainger Supplier No. 3WAJ3
This lightweight carboy from Dynalon Labware has large carrying handles and indented hand grips on the bottom for easy carrying and pouring. The high-density polyethylene (HDPE) material has high impact strength and excellent compatibility with weak and concentrated acids, bases, and alcohol. Supplied with a 63mm threaded closure that can be drilled to accept a faucet. Overall Height 342 mm, Overall Depth 245 mm, Overall Width248 mm