Learn More
Foxx Life Sciences EZBio® 75L (20 GAL) Gamma Sterilized Double Bagged HDPE Space Saving Carboy with Wide Mouth 120mm VersaCap® and 79.5L Max Capacity, Large Pinched Handels
Catalog No. 501720675
Encompass_Preferred
Foxx Life Sciences EZBio® Gamma Sterile HDPE Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. Assemblies are made in entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.HDPE carboys are made from a USP Class VI, FDA Grade materials.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options.
Catalog No. 50-172-0675 Supplier Foxx Life Sciences Supplier No. 5125K61FLS
Foxx Life Sciences EZBio® Gamma Sterile HDPE Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. Assemblies are made in entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.HDPE carboys are made from a USP Class VI, FDA Grade materials.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options.