Learn More
US Plastic Corporation CARBOY,POLYPROP,15L,WIDE MOUTH
SDP
Catalog No. NC2237196
Encompass
15 Liter Diamond® RealSeal™ Round Wide Mouth Polypropylene Carboy.Diamond® RealSeal™ Polypropylene Plastic Carboys meet the ample volume liquid storage and dispensing needs of the most demanding laboratory applications. Rugged and durable, these carboys are the safe, shatterproof alternative to glass for mixing, storing, and dispensing liquids
This item is not returnable. View return policy
Catalog No. NC2237196 Supplier US Plastic Corporation Supplier No. 84775
15 Liter Diamond® RealSeal™ Round Wide Mouth Polypropylene Carboy.Diamond® RealSeal™ Polypropylene Plastic Carboys meet the ample volume liquid storage and dispensing needs of the most demanding laboratory applications. Rugged and durable, these carboys are the safe, shatterproof alternative to glass for mixing, storing, and dispensing liquids