Learn More
Hach Company Carboy, Rectangular, with Spigot, 9 L
Supplier:  Hach Company 2098061
Encompass
Space-saving rectangular shape. With molded graduations. High-density polyethylene ensures tough, rigid carboy.20 L (5 gal) size has molded-in hand grips. Quick action spigot gives easy fluid control; incorporates two PTFE o-rings for positive seal.
Catalog No. 50-199-5374