Learn More
Medchemexpress LLC HY-W011651 25mg Medchemexpress, Carbutamide CAS:339-43-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01165125MG
Encompass_Preferred
Medchemexpress, HY-W011651 25mg Carbutamide CAS:339-43-5 Carbutamide (BZ-55) is an orally active and first-generation sulfonylurea with hypoglycemic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3316