Learn More
Cayman Chemical Carbutamide, Purity: ≥98%, CAS: 339-43-5, 50 mg
SDP
Catalog No. 502913717
Encompass_Preferred
A first generation sulfonylurea; inhibits KATP channels in β-cells (IC50 = 173 µM); reduces blood glucose levels in anesthetized dogs at 50 mg/kg; increases ventricular fibrillation in a rat model of LAD coronary artery ligation-induced cardiac ischemia
Catalog No. 50-291-3717 Supplier Cayman Chemical Supplier No. 3555250 MG
A first generation sulfonylurea; inhibits KATP channels in β-cells (IC50 = 173 µM); reduces blood glucose levels in anesthetized dogs at 50 mg/kg; increases ventricular fibrillation in a rat model of LAD coronary artery ligation-induced cardiac ischemia