Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carcainium chloride | 1042-42-8 | 99.5% | 347.84 g/mol | C18H22ClN3O2 | 50 MG
SDP

Catalog No. 5000242441
Encompass_Preferred

Carcainium chloride is a quaternary derivative of lidocaine used as a research chemical with reported antitussive effects. It is provided as a characterized solid and formulation-ready solutions with solubility and storage data to support assay and pharmacology studies.

  • Purity 99.47%.
  • Molecular weight 347.84 g/mol.
  • Soluble in DMSO (≈33.33 mg/mL) and compatible with formulated aqueous vehicles.
  • Available as solid and as 10 mM solution in DMSO.
  • Recommended storage 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Used in pharmacology and neuroscience research, including antitussive and histamine receptor studies.

Catalog No. 50-002-42441 Supplier Medchemexpress LLC Supplier No. HY106372A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carcainium chloride is a quaternary derivative of lidocaine used as a research chemical with reported antitussive effects. It is provided as a characterized solid and formulation-ready solutions with solubility and storage data to support assay and pharmacology studies.

  • Purity 99.47%.
  • Molecular weight 347.84 g/mol.
  • Soluble in DMSO (≈33.33 mg/mL) and compatible with formulated aqueous vehicles.
  • Available as solid and as 10 mM solution in DMSO.
  • Recommended storage 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Used in pharmacology and neuroscience research, including antitussive and histamine receptor studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.