Learn More
Medchemexpress LLC HY-106372A 10mg Medchemexpress, Carcainium (chloride) CAS:1042-42-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106372A10MG
Encompass_Preferred
Medchemexpress, HY-106372A 10mg Carcainium (chloride) CAS:1042-42-8 Carcainium chloride (QX 572) is a quaternary derivative of Lidocaine. Antitussive effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8339