Learn More
Cosmo Bio Usa Inc CARD HyperOva, 5 x 600ul
SDP
Catalog No. NC1651428
Encompass
CARD HyperOva is an ultra-superovulation reagent that induces ovulation at higher efficiency when compared with the conventional superovulation induction method (PMSG-hCG method). Following the conventional method, we were only able to obtain approximately 20 ova per mouse (C57BL/6J) on average; using CARD HyperOva, however, we can obtain as many as 3-4 times more ova.
Catalog No. NC1651428 Supplier Cosmo Bio Usa Inc Supplier No. KYD01006EXX5
CARD HyperOva is an ultra-superovulation reagent that induces ovulation at higher efficiency when compared with the conventional superovulation induction method (PMSG-hCG method). Following the conventional method, we were only able to obtain approximately 20 ova per mouse (C57BL/6J) on average; using CARD HyperOva, however, we can obtain as many as 3-4 times more ova.