Learn More
Mopec Card Mailer
SDP
Catalog No. 22446007 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
7.125 x 4.5 in.
Catalog No. Dimensions (L x W)
22-446-007 7.125 x 4.5 in.
1 options
Catalog No. 22-446-007 Supplier Mopec Supplier No. FS237
Add to Cart
Add to Cart
Secure packaging solution for the safe storage and transport of cards containing collected biological samples.
  • Designed for transporting FTX Matrix Cards.
  • Helps protect samples from contamination.
  • Shields samples from environmental exposure.
  • Helps prevent physical damage during storage and transport.
  • Maintains sample integrity for forensic analysis.
Specifications
Dimensions (L x W) 7.125 x 4.5 in.
Product Type Card Mailer
Color White
Quantity 50/Pk.