Learn More
Aviva Systems Biology Corp NLRC4 Antibody - C-terminal region (AVARP00019_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP00019_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NLRC4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NLRC4. Alternate names: CLAN, IPAF, CLAN1, CLANA, CLANB, CLANC, CLAND, CARD12, CLR2.1ApplicationIHC, WB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-62452