Learn More
Novus Biologicals™ CARD12 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 58484 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-78979. For further blocking peptide related protocol, click here . |
| Gene Alias | CARD12leucine rich repeat and CARD domaincontaining 4, caspase recruitment domain family, member 12, Caspase recruitment domain-containing protein 12, Clan protein, CLAN1ipaf, CLANCARD, LRR, and NACHT-containing protein, CLR2.1, Ice protease-activating factor, Ipaf, NLR family, CARD domain containing 4 |
| Gene Symbol | NLRC4 |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Immunology, Innate Immunity |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.