Learn More
Aviva Systems Biology Corp CARD9 Antibody - middle region (ARP57704_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57704_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CARD9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CARD9. Alternate names: hCARD9, CANDF2ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse
Catalog No. 50-286-354