Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cardamonin | 18956-16-6 | MFCD00238554 | 98.4% | 270.28 g/mol | C16H14O4 | 10 MG
SDP

Catalog No. 5000295503
Encompass_Preferred

Cardamonin is a chalcone-class flavonoid research compound supplied for laboratory use. It is provided as a 10 mg pack (C16H14O4, 270.28 g/mol) and is commonly used in cell-based and biochemical studies involving inflammatory and cancer signaling pathways. For research use only.

  • Purity of 98.4%.
  • Chemical formula C16H14O4 and molecular weight 270.28 g/mol.
  • Suitable for cell-based and biochemical assays.
  • Provided as a 10 mg pack for laboratory use.
  • Synonyms include cardamomin and alpinetin chalcone.

Catalog No. 50-002-95503 Supplier Medchemexpress LLC Supplier No. HYN027910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cardamonin is a chalcone-class flavonoid research compound supplied for laboratory use. It is provided as a 10 mg pack (C16H14O4, 270.28 g/mol) and is commonly used in cell-based and biochemical studies involving inflammatory and cancer signaling pathways. For research use only.

  • Purity of 98.4%.
  • Chemical formula C16H14O4 and molecular weight 270.28 g/mol.
  • Suitable for cell-based and biochemical assays.
  • Provided as a 10 mg pack for laboratory use.
  • Synonyms include cardamomin and alpinetin chalcone.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.