Learn More
Medchemexpress LLC HY-N0279 5mg Medchemexpress, Cardamonin CAS:18956-16-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02795MG
Encompass_Preferred
Medchemexpress, HY-N0279 5mg Cardamonin CAS:18956-16-6 Cardamonin (Cardamomin), a natural flavone isolated from Alpinia katsumadai Hayata, acts as an aryl hydrocarbon receptor (AhR) activator. Cardamonin alleviates inflammatory bowel disease by the inhibition of NLRP3 inflammasome activation via an AhR/Nrf2/NQO1 pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0247