Learn More
TARGETMOL CHEMICALS INC Cardanol C15 1 2MG
SDP
Catalog No. 502879653
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cardanol (C151), found in cashew nut shell liquid, induces mitochondria-associated apoptosis in human melanoma cells. Purity 98.48%
Catalog No. 50-287-9653 Supplier TARGETMOL CHEMICALS INC Supplier No. TN35942MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cardanol (C151), found in cashew nut shell liquid, induces mitochondria-associated apoptosis in human melanoma cells. Purity 98.48%