Learn More
Sarstedt Inc Cardboard box, 49 place capacity, 5x5x4in, 10/pack
Supplier:  Sarstedt Inc 95.064.949
Encompass_Preferred
Cryo box for sample storage. For 63-103mm height tumbes with max 17mm diameter.
Catalog No. NC0873135