Learn More
Alkali Scientific ASI™ Cardboard Cryo Freezer Box, 3.75" Boxes -Water and Ice Resistant Coating, w/16, 1/Pack
SDP
Catalog No. 503026879
Encompass_Preferred
Freezer boxes are protected from liquid nitrogen or condensation from freezers by a moisture repellent coating. Our freezer boxes are made of a sturdy white cardbard and fit all standard-stainless steel and polycarbonate storage systems. Small holes on the bottom allow for safe drainage of liquid nitrogen or water. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196. Made of sturdy White cardboard Moisture repellent coating Fits all standard stainless steel storage systems Drain holes in bottom corners 3 7/8″ White water repellent cardboard box for storage of samples or tubes. With Cell Dividers, with 4 drain holes Dimensions (L x W x H) 5 1/4 x 5 1/4 x 3 7/8″ ( 133 x 133 x 98mm)
Catalog No. 50-302-6879 Supplier Alkali Scientific Supplier No. FB3.75C16
Freezer boxes are protected from liquid nitrogen or condensation from freezers by a moisture repellent coating. Our freezer boxes are made of a sturdy white cardbard and fit all standard-stainless steel and polycarbonate storage systems. Small holes on the bottom allow for safe drainage of liquid nitrogen or water. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196. Made of sturdy White cardboard Moisture repellent coating Fits all standard stainless steel storage systems Drain holes in bottom corners 3 7/8″ White water repellent cardboard box for storage of samples or tubes. With Cell Dividers, with 4 drain holes Dimensions (L x W x H) 5 1/4 x 5 1/4 x 3 7/8″ ( 133 x 133 x 98mm)