Learn More
Cole-Parmer Argos Technologies PolarSafe Cardboard Freezer Box with Divider, 5 13/16" x 5 13/16" x 4 15/16"; 36-Place
Catalog No. 501960878 Shop All Cole-Parmer Products
Encompass_Preferred
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL. Boxes can be ordered with dividers for a complete box configuration or without dividers to customize your storage needs.
Catalog No. 50-196-0878 Supplier Cole-Parmer Supplier No. 2440977
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL. Boxes can be ordered with dividers for a complete box configuration or without dividers to customize your storage needs.