Learn More
Electron Microscopy Sciences Cardboard Freezer Box 2"
SDP
Catalog No. 501902289
Encompass_Preferred
For use with Stainless Steel Freezer Racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Cardboard boxes can be labelled with Cryo-Tags for easy identification. Use with stainless steel freezer racks.
Catalog No. 50-190-2289 Supplier Electron Microscopy Sciences Supplier No. 6161565
For use with Stainless Steel Freezer Racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Cardboard boxes can be labelled with Cryo-Tags for easy identification. Use with stainless steel freezer racks.