Learn More
Analytical Sales & Services Cardboard Micro Tube Boxes for 0.2mL Tubes, 169 Wells
Catalog No. 5000806187
Encompass
Safe micro tube storage\nThese low profile boxes accommodate standard 0.2mL micro tubes and vessels of a similar size. They come complete with well dividers.\n\n• Store in mechanical freezers or vapor phase LN2*\n• Will fit into most standard freezer racking\n• 169 wells in a 13 x 13 format\n\nQty/Pkg: 10
Catalog No. 50-008-06187 Supplier Analytical Sales & Services Supplier No. MTB169
Safe micro tube storage\nThese low profile boxes accommodate standard 0.2mL micro tubes and vessels of a similar size. They come complete with well dividers.\n\n• Store in mechanical freezers or vapor phase LN2*\n• Will fit into most standard freezer racking\n• 169 wells in a 13 x 13 format\n\nQty/Pkg: 10