Learn More
Analytical Sales & Services Cardboard Micro Tube Boxes for 0.5mL Tubes, 100 Wells
Catalog No. 5000805493
Encompass
Safe micro tube storage\nThese low profile boxes accommodate standard 0.5mL micro tubes and vessels of a similar size. They come complete with well dividers.\n\n• Store in mechanical freezers or vapor phase LN2*\n• Will fit into most standard freezer racking\n• 100 wells in a 10 x 10 format\n\nQty/Pkg: 100
Catalog No. 50-008-05493 Supplier Analytical Sales & Services Supplier No. MTB100
Safe micro tube storage\nThese low profile boxes accommodate standard 0.5mL micro tubes and vessels of a similar size. They come complete with well dividers.\n\n• Store in mechanical freezers or vapor phase LN2*\n• Will fit into most standard freezer racking\n• 100 wells in a 10 x 10 format\n\nQty/Pkg: 100